Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbutaline Sulfate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18607244 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-244 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-244 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T10621GM

missing translation for 'validMainframeMsg'

T1062, 23031-32-5, (C12H19NO3)2·H2SO4

Spectrum™ Chemical Terbutaline Sulfate, USP is used in the management of asthma symptoms as a reliever inhaler. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 23031-32-5
Molecular Formula C24H40N2O10S
Molecular Weight (g/mol) 548.65
InChI Key KFVSLSTULZVNPG-UHFFFAOYNA-N
IUPAC Name bis(5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol); sulfuric acid
SMILES OS(O)(=O)=O.CC(C)(C)NCC(O)C1=CC(O)=CC(O)=C1.CC(C)(C)NCC(O)C1=CC(O)=CC(O)=C1

Specifications

CAS Max % 1
Ignition Residue 0.002
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 1 g
Percent Purity 98 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.