Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbutaline hemisulfate salt, MP Biomedicals™

Catalog No. ICN15674701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15674701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15674701 Supplier MP Biomedicals, Inc Supplier No. 0215674701

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23031-32-5
Molecular Formula C24H40N2O10S
Molecular Weight (g/mol) 548.648
InChI Key KFVSLSTULZVNPG-UHFFFAOYSA-N
Synonym terbutaline sulfate, terbutaline hemisulfate, terbutaline hemisulfate salt, 2-t-butylamino-1-3,5-dihydroxyphenyl ethanol, brethine tn, dsstox_cid_25437, dsstox_rid_80878, dsstox_gsid_45437, 5-2-tert-butylamino-1-hydroxyethyl benzene-1,3-diol hemisulfate
PubChem CID 441334
IUPAC Name 5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol;sulfuric acid
SMILES CC(C)(C)NCC(C1=CC(=CC(=C1)O)O)O.CC(C)(C)NCC(C1=CC(=CC(=C1)O)O)O.OS(=O)(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 274.3
Physical Form Powder
Chemical Name or Material Terbutaline hemisulfate salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.