accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2306-91-4

H3COOCH3CH3

CAS RN 2306-91-4

IUPAC Name: 3-methylbutyl decanoate
Synonyms: 3-methylbutyl decanoate
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: XDOGFYDZGUDBQY-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OCCC(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results