accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23062-91-1

IUPAC Name: ethyl 17-methoxy-6-methyl-12-oxa-1,7-diazapentacyclo[11.8.0.0³,¹¹.0⁴,⁸.0¹⁴,¹⁹]henicosa-3,5,8,10,14(19),15,17-heptaene-5-carboxylate
Synonyms: ethyl 17-methoxy-6-methyl-12-oxa-1,7-diazapentacyclo[11.8.0.0³,¹¹.0⁴,⁸.0¹⁴,¹⁹]henicosa-3,5,8,10,14(19),15,17-heptaene-5-carboxylate
Molecular Weight (g/mol): 392.46
Molecular Formula: C23H24N2O4
InChi Key: VDDUJINYXKGZLV-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C(C)NC2=CC=C3OC4N(CCC5=C4C=CC(OC)=C5)CC3=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results