accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrBr

CAS RN 231606-27-2

IUPAC Name: 1,3-dibromo-5-dodecylbenzene
Synonyms: 1,3-dibromo-5-dodecylbenzene
Molecular Weight (g/mol): 404.23
Molecular Formula: C18H28Br2
InChi Key: LBSQLYHNNHYVBN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results