CAS RN 2323027-38-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Ceapin A7
Selective inhibitor of ATF6α; sensitizes cells to ER stress