accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHN

CAS RN 23328-87-2

IUPAC Name: 2-heptadecyl-1H-imidazole
Synonyms: 2-heptadecyl-1H-imidazole
Molecular Weight (g/mol): 306.54
Molecular Formula: C20H38N2
InChi Key: YTWBFUCJVWKCCK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC1=NC=CN1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)