accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NONF(R)O(R)OH(S)HO(R)HO

CAS RN 2341-22-2

IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one
Synonyms: 5-fluorocytidine
Molecular Weight (g/mol): 261.21
Molecular Formula: C9H12FN3O5
InChi Key: STRZQWQNZQMHQR-UAKXSSHOSA-N
SMILES: NC1=NC(=O)N(C=C1F)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results