Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluorocytidine 97.0+%, TCI America™
SDP

Catalog No. F05341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05341G Supplier TCI America Supplier No. F05341G
Add to Cart
Add to Cart

Specifications

CAS 2341-22-2
Melting Point 193°C
Color White
Molecular Formula C9H12FN3O5
MDL Number MFCD00210953
Quantity 1 g
Synonym 5-fluorocytidine, cytidine, 5-fluoro, unii-vor4x0d7wr, vor4x0d7wr, 4-amino-1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-fluoropyrimidin-2 1h-one, 3elc, fluc, pubchem6331, 5-fcr
InChI Key STRZQWQNZQMHQR-UAKXSSHOSA-N
SMILES NC1=NC(=O)N(C=C1F)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one
Molecular Weight (g/mol) 261.21
PubChem CID 16861
ChEBI CHEBI:38190
Formula Weight 261.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorocytidine
Show More Show Less
RTECSNumber HA3890000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.