accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2345-51-9

IUPAC Name: but-3-ynoic acid
Synonyms: 3-butynoic acid but-3-insaeure
Molecular Weight (g/mol): 84.07
Molecular Formula: C4H4O2
InChi Key: KKAHGSQLSTUDAV-UHFFFAOYSA-N
SMILES: OC(=O)CC#C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results