CAS RN 2353-45-9
Thermo Scientific Chemicals Fast Green FCF
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L Synonym: C.I. 42053 PubChem CID: 16887 IUPAC Name: disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Thermo Scientific Chemicals Fast Green FCF, pure, certified
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L Synonym: C.I. 42053 PubChem CID: 16887 IUPAC Name: disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Fast Green FCF, MP Biomedicals
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L PubChem CID: 16887 IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Food Green No. 3, TCI America™
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L PubChem CID: 16887 IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Fast Green FCF 85.0+%, TCI America™
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L PubChem CID: 16887 IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Fast Green FCF 85.0+%, TCI America™
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L PubChem CID: 16887 IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Fisher Science Education™ Fast Green Solution, 1%
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L PubChem CID: 16887 IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Fisher Scientific Fast Green FCF, Fisher BioReagents
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L Synonym: FD & C Green No. 3 PubChem CID: 16887 IUPAC Name: disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Fast Green FCF (Certified Biological Stain), Fisher Chemical
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L Synonym: FD&C Green No. 3 PubChem CID: 16887 IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Fast green FCF (C.I. 42053), For Microscopy, Certistain™, MilliporeSigma™
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Molecular Weight (g/mol): 808.84 MDL Number: MFCD00013053 InChI Key: XJBPDZVCYTYRIN-UHFFFAOYSA-L Synonym: C.I. 42053 PubChem CID: 16887 IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
Thermo Scientific Chemicals Fast Green FCF, Electrophoresis Reagent
CAS: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Synonym: C.I. 42053