Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Green FCF, Fisher BioReagents

Catalog No. BP12310 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP123-10 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP123-10

missing translation for 'validMainframeMsg'

Fast Green FCF is a protein stain for collagenous tissue.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37H34N2Na2O10S3
Molecular Weight (g/mol) 808.84
MDL Number MFCD00013053
InChI Key XJBPDZVCYTYRIN-UHFFFAOYSA-L
Synonym FD & C Green No. 3
PubChem CID 16887
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O

Specifications

Melting Point 290°C
Assay Percent Range ≥90.0 %
Merck Index 15, 3978
Identification Pass Test
Lambda Max 620 to 624nm
Formula Weight 808.85
Percent Purity ≥90.0%
Packaging Glass Bottle
Color Index No. 42053
Color Red
Quantity 10 g
Physical Form Powder
Chemical Name or Material Fast Green FCF
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.