Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fast Green FCF, Electrophoresis Reagent

Catalog No. AAJ6657022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6657022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6657022 Supplier Thermo Scientific Chemicals Supplier No. J6657022

missing translation for 'validMainframeMsg'

CAS: 2353-45-9 | C37H34N2Na2O10S3, C37H34N2Na2O10S3 | 808.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37H34N2Na2O10S3
Synonym C.I. 42053

Specifications

Content And Storage Ambient temperatures; Store under Nitrogen
Formula Weight 808.86
Melting Point ≈290°C (dec.)
Quantity 100 g
Sensitivity Hygroscopic
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.