CAS RN 2365033-54-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ FNIR-Tag, NHS
Near-infrared fluorescent dye for labeling of amines; supplied as NHS ester