CAS RN 2365417-91-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ SiRA 2
Fluorogen for SiRA light-up aptamer. Application: confocal microscopy, SRM (including STED)