Learn More
Description
Application: confocal microscopy, super resolution microscopy (SRM) techniques, including super-resolution stimulated emission depletion (STED). Cell permeable.
Properties and Photophysical Data: SiRA 2 is extremely bright, photostable and allows the visualization of mRNA expression in bacteria in no-wash, quencher-free, live-cell imaging experiments. Excitation and emission maxima (λ) are 649 nM and 662 nM, respectively; quantum yield = 0.98; extinction coefficient = 86,000 M-1cm-1; KD = 403 nM.
Reconstitute in 200 μL for a 0.5 mM stock solution. A typical working concentration for this product is 500 nM, see references provided for more details.
We recommend using 5mM Mg2+ (MgCl2 or MgSO4) in all buffers/media for maximum fluorescence intensity.
Specifications
Specifications
| Chemical Name or Material | (5-((2-(2-(2-Aminoethoxy)ethoxy)ethyl)carbamoyl)-2-(7-(dimethylamino)-3-(dimethyliminio)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)benzoate) |
| CAS | 2365417-91-8 |
| Quantity | 60 μg |
| Target | Light-up Aptamers |
| Molecular Formula | C33H42N4O5Si |
| Purity | 0.98 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
