accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NF

CAS RN 2369-18-8

IUPAC Name: 2-fluoro-3-methylpyridine
Synonyms: 2-fluoro-3-methylpyridine
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H6FN
InChi Key: PHGXUFPYCAWEHK-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results