CAS RN 2369979-67-7
CAS RN 2369979-67-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ JHU 37152
High affinity and highly potent hM3Dq and hM4Di DREADD agonist; blood brain barrier penetrant