accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2371-42-8

H3CH3CCH3CH3OH

CAS RN 2371-42-8

IUPAC Name: 1,2,7,7-tetramethylbicyclo[2.2.1]heptan-2-ol
Synonyms: 2-methylisoborneol
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: LFYXNXGVLGKVCJ-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(C)(O)C2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)