accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHNOONHNOO

CAS RN 2379-57-9

IUPAC Name: 6,7-dinitro-1,2,3,4-tetrahydroquinoxaline-2,3-dione
Synonyms: DNQX
Molecular Weight (g/mol): 252.14
Molecular Formula: C8H4N4O6
InChi Key: RWVIMCIPOAXUDG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=C2NC(=O)C(=O)NC2=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)