CAS RN 2382-80-1
CAS RN 2382-80-1
IUPAC Name:
ethyl (2S)-3-(1H-indol-3-yl)-2-(N-methylformamido)propanoate
Synonyms:
ethyl (2S)-3-(1H-indol-3-yl)-2-(N-methylformamido)propanoate
Molecular Weight (g/mol):
274.32
Molecular Formula:
C15H18N2O3
InChi Key:
LYXGYFZZVMODTB-AWEZNQCLSA-N
SMILES:
CCOC(=O)[C@H](CC1=CNC2=CC=CC=C12)N(C)C=O
1
–
1
of
1
results
1
N-Acetyl-L-tryptophan Ethyl Ester 98.0+%, TCI America™

CAS: 2382-80-1 Molecular Formula: C15H18N2O3 Molecular Weight (g/mol): 274.32 MDL Number: MFCD00005643 InChI Key: KQGQONPKSKUHHT-AWEZNQCLSA-N Synonym: Ac-Trp-OEt PubChem CID: 2724382 IUPAC Name: ethyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate SMILES: CCOC(=O)C(CC1=CNC2=CC=CC=C21)NC(=O)C