accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 239087-09-3

IUPAC Name: 1-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: LCVWAXYGYMZJER-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=CC(=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results