accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23981-80-8

IUPAC Name: 2-(6-methoxynaphthalen-2-yl)propanoic acid
Synonyms: naprosyn
Molecular Weight (g/mol): 230.26
Molecular Formula: C14H14O3
InChi Key: CMWTZPSULFXXJA-UHFFFAOYNA-N
SMILES: COC1=CC=C2C=C(C=CC2=C1)C(C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results