CAS RN 2399455-45-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Lenalidomide 4'-PEG1-azide
Cereblon ligand with PEG linker and terminal azide for onward chemistry