accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 23996-12-5

IUPAC Name: 3-(2-phenyl-1H-imidazol-1-yl)propanenitrile
Synonyms: 3-(2-phenylimidazol-1-yl)propanenitrile
Molecular Weight (g/mol): 197.24
Molecular Formula: C12H11N3
InChi Key: BVYPJEBKDLFIDL-UHFFFAOYSA-N
SMILES: N#CCCN1C=CN=C1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)