Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Phenyl-1H-imidazol-1-yl)propanenitrile, Tech., Thermo Scientific™

Catalog No. MO01011EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01011EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01011EA Supplier Thermo Scientific Chemicals Supplier No. MO01011EA

Chemical Identifiers

CAS 23996-12-5
Molecular Formula C12H11N3
Molecular Weight (g/mol) 197.24
MDL Number MFCD00047047
InChI Key BVYPJEBKDLFIDL-UHFFFAOYSA-N
Synonym 1-2-cyanoethyl-2-phenylimidazole, 3-2-phenyl-1h-imidazol-1-yl propanenitrile, 1h-imidazole-1-propanenitrile, 2-phenyl, 2-phenyl-1h-imidazole-1-propiononitrile, 3-2-phenylimidazol-1-yl propanenitrile, 1h-imidazole-1-propanenitrile,2-phenyl, 3-2-phenyl-imidazol-1-yl-propionitrile, acmc-1cmrx, 2-phenyl-1h-imidazole-1-propanenitrile, 3-2-phenyl-1-imidazolyl propionitrile
PubChem CID 90325
IUPAC Name 3-(2-phenyl-1H-imidazol-1-yl)propanenitrile
SMILES N#CCCN1C=CN=C1C1=CC=CC=C1

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 197.24
Grade Technical
Chemical Name or Material 3-(2-Phenyl-1H-imidazol-1-yl)propanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.