Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Cyanoethyl)-2-phenylimidazole 98.0+%, TCI America™
SDP

Catalog No. C196425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C196425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C196425G Supplier TCI America Supplier No. C196425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23996-12-5
Molecular Formula C12H11N3
Molecular Weight (g/mol) 197.24
MDL Number MFCD00047047
InChI Key BVYPJEBKDLFIDL-UHFFFAOYSA-N
Synonym 1-2-cyanoethyl-2-phenylimidazole, 3-2-phenyl-1h-imidazol-1-yl propanenitrile, 1h-imidazole-1-propanenitrile, 2-phenyl, 2-phenyl-1h-imidazole-1-propiononitrile, 3-2-phenylimidazol-1-yl propanenitrile, 1h-imidazole-1-propanenitrile,2-phenyl, 3-2-phenyl-imidazol-1-yl-propionitrile, acmc-1cmrx, 2-phenyl-1h-imidazole-1-propanenitrile, 3-2-phenyl-1-imidazolyl propionitrile
PubChem CID 90325
IUPAC Name 3-(2-phenyl-1H-imidazol-1-yl)propanenitrile
SMILES N#CCCN1C=CN=C1C1=CC=CC=C1

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 25 g
Formula Weight 197.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Cyanoethyl)-2-phenylimidazole
EINECSNumber (5)-0412
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.