accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24157-81-1

IUPAC Name: 2,6-bis(propan-2-yl)naphthalene
Synonyms: DIPN
Molecular Weight (g/mol): 212.34
Molecular Formula: C16H20
InChi Key: GWLLTEXUIOFAFE-UHFFFAOYSA-N
SMILES: CC(C)C1=CC2=CC=C(C=C2C=C1)C(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results