accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClSNOO

CAS RN 2425-06-1

IUPAC Name: 2-[(1,1,2,2-tetrachloroethyl)sulfanyl]-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione
Synonyms: crisfolatan captafol (cis isomer) captafol (isomer unspecified)
Molecular Weight (g/mol): 349.05
Molecular Formula: C10H9Cl4NO2S
InChi Key: JHRWWRDRBPCWTF-UHFFFAOYNA-N
SMILES: ClC(Cl)C(Cl)(Cl)SN1C(=O)C2CC=CCC2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results