accessibility menu, dialog, popup

UserName

Viewing profile as user:
S

CAS RN 243-46-9

IUPAC Name: 17-thiatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),2,4,6,8,11(16),12,14-octaene
Synonyms: 17-thiatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),2,4,6,8,11(16),12,14-octaene
Molecular Weight (g/mol): 234.32
Molecular Formula: C16H10S
InChi Key: UWMISBRPSJFHIR-UHFFFAOYSA-N
SMILES: S1C2=C(C=CC=C2)C2=C1C=C1C=CC=CC1=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results