accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOCH3

CAS RN 24317-94-0

IUPAC Name: ethyl 2-acetylheptanoate
Synonyms: ethyl 2-acetylheptanoate
Molecular Weight (g/mol): 200.28
Molecular Formula: C11H20O3
InChi Key: XIGZBCUFFUBWDM-UHFFFAOYNA-N
SMILES: CCCCCC(C(C)=O)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results