accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24332-20-5

IUPAC Name: 1,1,2-trimethoxyethane
Synonyms: bis-methoxyethoxy methane
Molecular Weight (g/mol): 120.15
Molecular Formula: C5H12O3
InChi Key: DYOZNCVZPFIXLU-UHFFFAOYSA-N
SMILES: COCC(OC)OC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)