Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2-Trimethoxyethane 98.0+%, TCI America™
SDP

Catalog No. T083125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T083125ML 25 mL
T0831100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T083125ML Supplier TCI America Supplier No. T083125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24332-20-5
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00008486
InChI Key DYOZNCVZPFIXLU-UHFFFAOYSA-N
Synonym Methoxyacetaldehyde Dimethyl Acetal
PubChem CID 47520
IUPAC Name 1,1,2-trimethoxyethane
SMILES COCC(OC)OC

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 mL
UN Number 3271
Formula Weight 120.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2-Trimethoxyethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.