accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2434-53-9

CH3NOHNONH2

CAS RN 2434-53-9

IUPAC Name: 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-amino-1-methyl-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 141.13
Molecular Formula: C5H7N3O2
InChi Key: GZLZRPNUDBIQBM-UHFFFAOYSA-N
SMILES: CN1C(=O)NC(=O)C=C1N
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)