Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-1-methyluracil 98.0+%, TCI America™
SDP

Catalog No. A24495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24495G Supplier TCI America Supplier No. A24495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2434-53-9
Molecular Formula C5H7N3O2
Molecular Weight (g/mol) 141.13
MDL Number MFCD00075366
InChI Key GZLZRPNUDBIQBM-UHFFFAOYSA-N
Synonym 6-amino-1-methyluracil, 6-amino-1-methylpyrimidine-2,4 1h,3h-dione, 1-methyl-6-aminouracil, 2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl, 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-amino-1-methyl-2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl-1h-pyrimidine-2,4-dione, 6-amino-1-methyl-1,3-dihydropyrimidine-2,4-dione, 6-amino-1-methyl-3h-pyrimidine-2,4-dione, zlchem 437
PubChem CID 75520
IUPAC Name 6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CN1C(=O)NC(=O)C=C1N

Specifications

Melting Point 307°C
Color White
Quantity 5 g
Formula Weight 141.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-methyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.