accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHOHNNHSOOOH

CAS RN 244192-94-7

IUPAC Name: N-{[3-(3-{[2-(4-benzenesulfonamidophenyl)ethyl]amino}-2-hydroxypropoxy)phenyl]methyl}acetamide
Synonyms: N-{[3-(3-{[2-(4-benzenesulfonamidophenyl)ethyl]amino}-2-hydroxypropoxy)phenyl]methyl}acetamide
Molecular Weight (g/mol): 497.61
Molecular Formula: C26H31N3O5S
InChi Key: AWIONHVPTYTSHZ-UHFFFAOYNA-N
SMILES: CC(=O)NCC1=CC(OCC(O)CNCCC2=CC=C(NS(=O)(=O)C3=CC=CC=C3)C=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)