accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3

CAS RN 2445-78-5

IUPAC Name: 2-methylbutyl 2-methylbutanoate
Synonyms: 2-methylbutyl 2-methylbutanoate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: PVYFCGRBIREQLL-UHFFFAOYNA-N
SMILES: CCC(C)COC(=O)C(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results