accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 2446-69-7

IUPAC Name: 4-hexylphenol
Synonyms: 4-n-hexylphenol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: SZWBRVPZWJYIHI-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results