accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 2456-28-2

IUPAC Name: 1-(decyloxy)decane
Synonyms: didecyl ether
Molecular Weight (g/mol): 298.56
Molecular Formula: C20H42O
InChi Key: LTSWUFKUZPPYEG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results