Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decyl Ether 95.0+%, TCI America™
SDP

Catalog No. D003525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D003525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D003525ML Supplier TCI America Supplier No. D003525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2456-28-2
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.56
MDL Number MFCD00026539
InChI Key LTSWUFKUZPPYEG-UHFFFAOYSA-N
Synonym Didecyl Ether
PubChem CID 17152
IUPAC Name 1-(decyloxy)decane
SMILES CCCCCCCCCCOCCCCCCCCCC

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 mL
Formula Weight 298.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Decyl Ether
EINECSNumber (2)-3075
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.