accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNCl

CAS RN 2457-47-8

IUPAC Name: 3,5-dichloropyridine
Synonyms: 3,5-dichloropyridine
Molecular Weight (g/mol): 147.99
Molecular Formula: C5H3Cl2N
InChi Key: WPGHPGAUFIJVJF-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=CN=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)