CAS RN 24597-12-4
CAS RN 24597-12-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
281.24
Molecular Formula:
Cl4Ga2
InChi Key:
ZAIGZEHFDHYDNG-UHFFFAOYSA-N
SMILES:
[Cl].[Cl].[Cl].[Cl].[Ga].[Ga+3]
1
–
1
of
1
results
1
Gallium(II) chloride, ultra dry, 99.999% (metals basis)
CAS: 24597-12-4 MDL Number: MFCD00151526