accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOCH3H3CCH3

CAS RN 2460-77-7

IUPAC Name: 2,5-di-tert-butylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2,5-di-tert-butylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: ZZYASVWWDLJXIM-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)