Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Di-tert-butyl-1,4-benzoquinone 98.0+%, TCI America™
SDP

Catalog No. D017825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D017825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D017825G Supplier TCI America Supplier No. D017825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2460-77-7
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00019442
InChI Key ZZYASVWWDLJXIM-UHFFFAOYSA-N
Synonym 2,5-di-tert-butyl-1,4-benzoquinone, 2,5-di-tert-butylquinone, 2,5-di-tert-butylsemiquinone, 2,5-di-tert-butylbenzoquinone, 2,5-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,5-di-tert-butyl-p-quinone, 2,5-di-tert-butyl-p-benzoquinone, unii-g893osv02d, p-benzoquinone, 2,5-di-tert-butyl, 2,5-di-t-butyl-p-benzoquinone
PubChem CID 17161
IUPAC Name 2,5-ditert-butylcyclohexa-2,5-diene-1,4-dione
SMILES CC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)C

Specifications

Melting Point 153°C
Quantity 25 g
Formula Weight 220.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di-tert-butyl-1,4-benzoquinone
RTECSNumber GU5140000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.