accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNH2SOOOSOOO

CAS RN 24605-36-5

IUPAC Name: disodium 2-aminobenzene-1,4-disulfonate
Synonyms: disodium 2-aminobenzene-1,4-disulfonate
Molecular Weight (g/mol): 297.21
Molecular Formula: C6H5NNa2O6S2
InChi Key: QYUDDCLXBAJUNA-UHFFFAOYSA-L
SMILES: [Na+].[Na+].NC1=CC(=CC=C1S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results