Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline-2,5-disulfonic Acid Monosodium Salt 98.0+%, TCI America™
SDP

Catalog No. A0263500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0263500G 500 g
A026325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0263500G Supplier TCI America Supplier No. A0263500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24605-36-5
Molecular Formula C6H5NNa2O6S2
Molecular Weight (g/mol) 297.21
MDL Number MFCD00078393
InChI Key QYUDDCLXBAJUNA-UHFFFAOYSA-L
Synonym 2-Amino-1,4-benzenedisulfonic Acid Monosodium Salt, 2,5-Disulfoaniline Monosodium Salt, Sodium Hydrogen 2-Amino-1,4-benzenedisulfonate
PubChem CID 87094165
IUPAC Name disodium 2-aminobenzene-1,4-disulfonate
SMILES [Na+].[Na+].NC1=CC(=CC=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 275.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Aniline-2,5-disulfonic Acid Monosodium Salt
EINECSNumber (3)-2101&(3)-3169
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.