CAS RN 2463-77-6
MP Biomedicals, Inc 2-Undecen-1-al, MP Biomedicals
CAS: 2463-77-6 Molecular Formula: C11H20O Molecular Weight (g/mol): 168.28 InChI Key: PANBRUWVURLWGY-MDZDMXLPSA-N Synonym: trans-2-undecenal,2-undecenal,e-undec-2-enal,e-2-undecenal,2-undecenal, 2e,2-undecen-1-al,2e-2-undecenal,trans-2-undecen-1-al,2-undecenal, e,unii-fnp3s9mg30 PubChem CID: 5283356 IUPAC Name: (E)-undec-2-enal SMILES: CCCCCCCCC=CC=O