accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24667-21-8

IUPAC Name: 2-(2-aminoacetamido)-3-carbamoylpropanoic acid
Synonyms: 2-(2-aminoacetamido)-3-carbamoylpropanoic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C6H11N3O4
InChi Key: FUESBOMYALLFNI-UHFFFAOYNA-N
SMILES: NCC(=O)NC(CC(N)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results