accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 246852-46-0

IUPAC Name: 1-[2-(cycloheptyloxy)-2-(2,6-dichlorophenyl)ethenyl]-1H-1,2,4-triazole
Synonyms: 1-[2-(cycloheptyloxy)-2-(2,6-dichlorophenyl)ethenyl]-1,2,4-triazole
Molecular Weight (g/mol): 352.26
Molecular Formula: C17H19Cl2N3O
InChi Key: STCVFKBRXOEQRF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1C(OC1CCCCCC1)=CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results