accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3COH2COSiCH3CH3H3C

CAS RN 24697-35-6

IUPAC Name: 2,2,6,6-tetramethyl-4-methylidene-3,5-dioxa-2,6-disilaheptane
Synonyms: 2,2,6,6-tetramethyl-4-methylidene-3,5-dioxa-2,6-disilaheptane
Molecular Weight (g/mol): 204.42
Molecular Formula: C8H20O2Si2
InChi Key: QTLOQPVMBORRRD-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=C)O[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results